Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Thermo Scientific Chemicals

Phosphotungstic acid hydrate

CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 208311000
Price (USD)
187.65
Special offer
Online exclusive
Ends: 30-Jun-2026
221.00
Save 33.35 (15%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
187.65
Special offer
Online exclusive
Ends: 30-Jun-2026
221.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS12501-23-4
IUPAC Name12-Tungstophosphate hydrate
Molecular Weight (g/mol)2880.20
MDL NumberMFCD00011342
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<50 ppm
Sodium (Na)=<100 ppm
Appearance (Color)White to yellow to beige to grey
Assay>=82 % (WO3)
Chloride (Cl)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only