Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide
Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide
Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide
Acros Organics

Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide

CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic drum
Catalog number 211070010
HSN 28530099
Price (INR)
8,622.00
Each
Quantity:
1 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (INR)
8,622.00
Each
Chemical Identifiers
CAS39409-82-0
IUPAC Namepentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular FormulaC4H10Mg5O18
InChI KeyYMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILESO.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<700 ppm
Sulfate (SO4)=<3000 ppm
Arsenic (As)=<2 ppm
Calcium (Ca)=<7500 ppm
Iron (Fe)=<400 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only