Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Acros Organics

Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 211570010
also known as 21157
HSN 29152990
Price (INR)
14,108.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
14,108.00
Each
Chemical Identifiers
CAS16674-78-5
IUPAC Namemagnesium(2+) diacetate tetrahydrate
Molecular FormulaC4H14MgO8
InChI KeyXKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILESO.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<500 ppm
Infrared spectrumConforms
Chloride (Cl)=<100 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only