Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure, 60mL - FAQs
Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure, 60mL - FAQs
View additional product information for Nalgene™ Wide-Mouth Straight-Sided PPCO Jars with Closure - FAQs (2118-0001, 2118-0002, 2118-0004, 2118-0016, 2118-0008, 2118-0032, 2118-0001PK, 2118-0032PK, 2118-0002PK, 2118-9050, 2118-0004PK, 2118-0016PK, 2118-9050PK, 2118-0008PK)
2 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Why are some of your wide mouth bottles and jars not considered liquid leakproof?
This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.