Mercury(I) chloride, 99+%, extra pure
Mercury(I) chloride, 99+%, extra pure
Mercury(I) chloride, 99+%, extra pure
Thermo Scientific Chemicals

Mercury(I) chloride, 99+%, extra pure

CAS: 10112-91-1 | Cl2Hg2 | 472.08 g/mol
数量:
5 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 213125000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10112-91-1
IUPAC Namedimercuranebis(ylium) dichloride
Molecular FormulaCl2Hg2
InChI KeyZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg+][Hg+]
さらに表示
Appearance (Form)Powder
Residue after reduction=<0.02 %
Mercuric Chloride=<0.01 % (HgCl2)
Appearance (Color)Off-white
Titration Iodimetric>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only