1-Hexene, 97%
1-Hexene, 97%
1-Hexene, 97%
Thermo Scientific Chemicals

1-Hexene, 97%

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15 | CAS: 592-41-6 | C6H12 | 84.15 g/mol
数量:
100 mL
2.5 L
25 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 213210025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
さらに表示
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3867 to 1.3887 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only