Sodium carbonate decahydrate, 99+%, for analysis
Sodium carbonate decahydrate, 99+%, for analysis
Sodium carbonate decahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, for analysis

CAS: 6132-02-1 | CNa2O3·10H2O | 286.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 213500010
Price (USD)
89.60
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
89.60
Each
Chemical Identifiers
CAS6132-02-1
IUPAC Namedisodium decahydrate carbonate
Molecular FormulaCH20Na2O13
InChI KeyXYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl>=99.0 %
Heavy metals (as Pb)=<10 ppm
Total nitrogen (as N)=<0.0005 %
Appearance (Color)White
PO4 and SiO2 (as SiO2)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only