Difco™ Casein Digest
Gibco™

Difco™ Casein Digest

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein.
製品番号(カタログ番号)数量
21489510 kg
2148962 kg
製品番号(カタログ番号) 214895
価格(JPY)
-
数量:
10 kg

Difco Casein Digest is an animal origin (AO) enzymatic digest of casein with different enzymatic digestion conditions than other casein digests in our portfolio, such as Bacto Casitone and Bacto Tryptone. Difco Casein Digest has a unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites ideal for growth promotion in fermentation applications, especially Escherichia coli.

Features:

  • Unique nutritional profile of amino acids, nutrients, minerals, nucleotide precursors, peptides, and other metabolites

Applications of Difco Casein Digest include:

  • Microbial fermentation
  • Growth promotion, specifically E. Coli

Applications:

Cultivation of heterotrophic microorganisms

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
仕様
添加剤Casein Digest
認証/コンプライアンスLUO, MUO
Light Beige
使用対象(アプリケーション)Cultivation of heterotrophic microorganisms
フォーマットPail
グレードTechnical
培地タイプCell Culture Supplement
製品ラインDifco
数量10 kg
品質保持期間1825 Days
形状Free-flowing Powder
製造元United States
製品タイプPeptone
無菌性Non-sterile
Unit Size10 kg
組成および保存条件
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.