Tetracosanoic acid, 99%
Tetracosanoic acid, 99%
Tetracosanoic acid, 99%
Thermo Scientific Chemicals

Tetracosanoic acid, 99%

CAS: 557-59-5 | C24H48O2 | 368.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 215640010
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS557-59-5
IUPAC Nametetracosanoic acid
Molecular FormulaC24H48O2
InChI KeyQZZGJDVWLFXDLK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes
Infrared spectrumConforms
Appearance (Color)White
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only