3-(Trimethoxysilyl)propyl methacrylate, 98%
3-(Trimethoxysilyl)propyl methacrylate, 98%
3-(Trimethoxysilyl)propyl methacrylate, 98%
Thermo Scientific Chemicals

3-(Trimethoxysilyl)propyl methacrylate, 98%

CAS: 2530-85-0 | C10H20O5Si | 248.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 216555000
Price (USD)
79.90
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.90
Each
Chemical Identifiers
CAS2530-85-0
IUPAC Name3-(trimethoxysilyl)propyl 2-methylprop-2-enoate
Molecular FormulaC10H20O5Si
InChI KeyXDLMVUHYZWKMMD-UHFFFAOYSA-N
SMILESCO[Si](CCCOC(=O)C(C)=C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<70 APHA
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only