L(+)-alpha-Phenylglycine, 99%
L(+)-alpha-Phenylglycine, 99%
L(+)-alpha-Phenylglycine, 99%
Thermo Scientific Chemicals

L(+)-alpha-Phenylglycine, 99%

CAS: 2935-35-5 | C8H9NO2 | 151.16 g/mol
数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 217991000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2935-35-5
IUPAC Name2-amino-2-phenylacetic acid
Molecular FormulaC8H9NO2
InChI KeyZGUNAGUHMKGQNY-UHFFFAOYNA-N
SMILESNC(C(O)=O)C1=CC=CC=C1
さらに表示
Loss on drying=<0.5 % (1 g, 105°C, 3 h)
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=98.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only