Trimethyl orthoacetate, 98+%
Trimethyl orthoacetate, 98+%
Trimethyl orthoacetate, 98+%
Thermo Scientific Chemicals

Trimethyl orthoacetate, 98+%

CAS: 1445-45-0 | C5H12O3 | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 218181000
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.10
Save 8.45
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.10
Save 8.45
Each
Chemical Identifiers
CAS1445-45-0
IUPAC Name1,1,1-trimethoxyethane
Molecular FormulaC5H12O3
InChI KeyHDPNBNXLBDFELL-UHFFFAOYSA-N
SMILESCOC(C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Refractive index1.3870 to 1.3900 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only