Aluminum nitrate nonahydrate, 99+%, for analysis
Aluminum nitrate nonahydrate, 99+%, for analysis
Aluminum nitrate nonahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Aluminum nitrate nonahydrate, 99+%, for analysis

CAS: 7784-27-2 | AlN3O9·9H2O | 375.13 g/mol
数量:
100 g
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 218281000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7784-27-2
IUPAC Namealuminium(3+) nonahydrate trinitrate
Molecular FormulaAlH18N3O18
InChI KeySWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Titration Complexometric>=99.0 %
pH2.5 to 3.5 (5 % at 20°C)
Appearance (Color)Semitransparent to transparent
Heavy metals (as Pb)=<20 ppm
Sulfate (SO4)=<50 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only