Thermo Scientific Chemicals

Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 218322500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 218322500
250 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
10377-48-7
IUPAC Name
dilithium(1+) sulfate
Molecular Formula
Li2O4S
InChI Key
INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]S([O-])(=O)=O
Water
=<1 % (K.F.)
Appearance (Form)
Powder
Insoluble matter
=<0.01 %
Appearance (Color)
White
Titration after Ion exchange
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text