N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
N,N-Dimethylethylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylethylamine, 99%

CAS: 598-56-1 | C4H11N | 73.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 219290010
Price (EUR)
117,65
Online exclusive
130,00
Save 12,35 (10%)
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
117,65
Online exclusive
130,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS598-56-1
IUPAC Nameethyldimethylamine
Molecular FormulaC4H11N
InChI KeyDAZXVJBJRMWXJP-UHFFFAOYSA-N
SMILESCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.3715 to 1.3735 (20°C, 589 nm)
Appearance (Form)Liquid
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only