Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Thermo Scientific Chemicals

Triethylamine, 99.7%, extra pure

CAS: 121-44-8 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
1 kg
2.5 kg
10 kg
Catalog number 219510500
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.40
Save 13.75
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 28-Dec-2026
91.40
Save 13.75
Each
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.6 %
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.10 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only