Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 220151000
100 g, Each
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 220151000
100 g, Each
Quantity
Catalog number: 220151000
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
127-08-2
IUPAC Name
potassium acetate
Molecular Formula
C2H3KO2
InChI Key
SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES
[K+].CC([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 %
Appearance (Form)
Crystalline or granular powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text