Hexicarbonilo de cromo, +99 %, Thermo Scientific Chemicals
Hexicarbonilo de cromo, +99 %, Thermo Scientific Chemicals
Hexicarbonilo de cromo, +99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexicarbonilo de cromo, +99 %, Thermo Scientific Chemicals

CAS: 13007-92-6 | C6CrO6 | 220.06 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
10 g
50 g
Envase:
Frasco de vidrio
Número de catálogo 221050500
Precio (USD)
-
Cantidad:
50 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS13007-92-6
IUPAC Namehexakis(methanidylidyneoxidanium) chromium
Molecular FormulaC6CrO6
InChI KeyKOTQLLUQLXWWDK-UHFFFAOYSA-N
SMILES[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to yellow
Infrared spectrumConforms
Elemental Analysis>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only