Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Lead(II,IV) oxide, red, 98%
Thermo Scientific Chemicals

Lead(II,IV) oxide, red, 98%

CAS: 1314-41-6 | O4Pb3 | 685.57 g/mol
数量:
100 g
500 g
5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 221115000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1314-41-6
IUPAC Namebicyclo[3.1.1]triplumboxan-1-yl
Molecular FormulaO4Pb3
InChI KeyXMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILESO1[Pb]2O[Pb]1O[Pb]O2
さらに表示
Calcium (Ca)=<100 ppm
Iron (Fe)=<50 ppm
Sodium (Na)=<100 ppm
Potassium (K)=<100 ppm
Zinc (Zn)=<50 ppm
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only