1,1,3,3-Tetramethylguanidine, 99%
1,1,3,3-Tetramethylguanidine, 99%
1,1,3,3-Tetramethylguanidine, 99%
Thermo Scientific Chemicals

1,1,3,3-Tetramethylguanidine, 99%

CAS: 80-70-6 | C5H13N3 | 115.18 g/mol
数量:
100 g
500 g
2 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 221290020
価格(JPY)
-
数量:
2 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS80-70-6
IUPAC NameN,N,N',N'-tetramethylguanidine
Molecular FormulaC5H13N3
InChI KeyKYVBNYUBXIEUFW-UHFFFAOYSA-N
SMILESCN(C)C(=N)N(C)C
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.4665 to 1.4695 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only