Benzoic acid, 99.5%, for analysis
Benzoic acid, 99.5%, for analysis
Benzoic acid, 99.5%, for analysis
Thermo Scientific Chemicals

Benzoic acid, 99.5%, for analysis

CAS: 65-85-0 | C7H6O2 | 122.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 221800010
Price (USD)
173.65
Online exclusive
193.00
Save 19.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
173.65
Online exclusive
193.00
Save 19.35
Each
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point121°C to 123°C
Sulfated ash=<0.01 %
Iron (Fe)=<4 ppm
Infrared spectrumConforms
Titration with NaOH>=99.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only