Powder Rheology Accessories for HAAKE MARS Rheometers
Thermo Scientific™
Powder Rheology Accessories for HAAKE MARS Rheometers
Enhance powder analysis with Thermo Scientific HAAKE MARS Rheometers. Measure flowability, compressibility, and shear properties with precision and ease.
Have Questions?
Catalog NumberType
222-2632Rheometer Accessory Kit
Catalog number 222-2632
Price (USD)
-
Type:
Rheometer Accessory Kit
The Thermo Scientific Powder Rheology Accessories for Thermo Scientific HAAKE MARS Rheometers enable precise measurement of powder flowability, compressibility, and shear properties. These accessories are designed to enhance product consistency, reduce waste, and speed up development cycles. With the flexible Powder Flow Cell, user-friendly RheoWin Software, and predefined test protocols, users can achieve reliable and reproducible results across various applications, including pharmaceuticals, food processing, and battery manufacturing.
The Thermo Scientific Powder Rheology Accessories for Thermo Scientific HAAKE MARS Rheometers provide a comprehensive solution for analyzing powder flowability, compressability, and other powder properties. Designed for versatility and precision, these accessories support a wide range of applications and deliver valuable insights into powder behavior.
  • Flexible Powder Flow Cell: Enhances measurement precision across various applications, from pharmaceuticals to food processing.
  • User-friendly RheoWin Software: Ensures easy handling and customization, saving time and resources.
  • Predefined Test Protocols: Allows efficient, standardized, or custom measurements.
  • Advanced Measurement Capabilities: Measure Basic Flowability Energy (BFE), Stability Index (SI), Major Principal Stress (MPS), and Unconfined Yield Stress (UYS).
  • Touchscreen Operation: Conveniently runs Standard Operating Procedures (SOPs) directly on the device's touchscreen.
  • Safety Features: The 'Protect Assist' safety door links measurement sequences to the door status, ensuring safe and secure testing.
  • Comprehensive Toolkit: Includes all necessary components for compression, powder flow, and powder shear tests, delivered in a high-quality carrying case.
Specifications
For Use With (Equipment)HAAKE MARS Rheometers
Includes
  • AM-PD-C Adapter module 
  • Powder filling tool with powder filling funnel
  • Powder cup CCB28 ST-PD
  • Powder cup CCB28 PD 
  • Twisted blade rotor FL27 2B-PD/SS
  • Rotor P28 PO-PD/SS for compression test
  • P28 ST-PD/SS for powder shear test
  • Two parts powder collector
  • Powder cup holder
  • Powder scoop
  • HAAKE RheoWin software module 
  • Safety door 'Protect Assist'
TypeRheometer Accessory Kit
Unit SizeEach