Thermo Scientific Chemicals

Ammonium sodium phosphate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 222640025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium sodium phosphate tetrahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 222640025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222640025
Price (USD)
Price: 137.00
Your price:
Quantity
-

Chemical Identifiers

CAS
7783-13-3
IUPAC Name
ammonium sodium tetrahydrate hydrogen phosphate
Molecular Formula
H13NNaO8P
InChI Key
IQTQISLCLWJRPM-UHFFFAOYSA-M
SMILES
[NH4+].O.O.O.O.[Na+].OP([O-])([O-])=O
Lead (Pb)
=<10 ppm
Appearance (Form)
Crystalline powder or crystals
Insoluble matter
=<0.01 % (in NH4OH)
Nitrate (NO3)
=<20 ppm
Arsenic (As)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text