Nickel(II) acetate tetrahydrate, 99+%, for analysis
Nickel(II) acetate tetrahydrate, 99+%, for analysis
Nickel(II) acetate tetrahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 99+%, for analysis

CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 223140025
Price (USD)
249.65
Online exclusive
277.00
Save 27.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
249.65
Online exclusive
277.00
Save 27.35
Each
Chemical Identifiers
CAS6018-89-9
IUPAC Namenickel(2+) diacetate tetrahydrate
Molecular FormulaC4H14NiO8
InChI KeyOINIXPNQKAZCRL-UHFFFAOYSA-L
SMILESO.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.0 %
Aluminium (Al)=<5 ppm
Calcium (Ca)=<100 ppm
Cadmium (Cd)=<50 ppm
Cobalt (Co)=<2000 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only