Tetrakis(triphenylphosphine)platinum(0), 98%
Tetrakis(triphenylphosphine)platinum(0), 98%
Tetrakis(triphenylphosphine)platinum(0), 98%
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)platinum(0), 98%

CAS: 14221-02-4 | C72H60P4Pt | 1244.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 223960010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14221-02-4
IUPAC Nametetrakis(triphenylphosphane) platinum
Molecular FormulaC72H60P4Pt
InChI KeySYKXNRFLNZUGAJ-UHFFFAOYSA-N
SMILES[Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to light brown
Carbon (C)67.8 to 71.2 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only