Methyltriphenylphosphonium iodide, 97%
Methyltriphenylphosphonium iodide, 97%
Methyltriphenylphosphonium iodide, 97%
Thermo Scientific Chemicals

Methyltriphenylphosphonium iodide, 97%

CAS: 2065-66-9 | C19H18IP | 404.23 g/mol
数量:
25 g
100 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 224031000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2065-66-9
IUPAC Namemethyltriphenylphosphanium iodide
Molecular FormulaC19H18IP
InChI KeyJNMIXMFEVJHFNY-UHFFFAOYSA-M
SMILES[I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Form)Powder
Melting point183°C to 188°C
Titration Argentometric>=96.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only