N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
Thermo Scientific Chemicals

N,N-Dimethyloctylamine, 97%

CAS: 7378-99-6 | C10H23N | 157.30 g/mol
数量:
25 mL
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 225295000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7378-99-6
IUPAC Namedimethyl(octyl)amine
Molecular FormulaC10H23N
InChI KeyUQKAOOAFEFCDGT-UHFFFAOYSA-N
SMILESCCCCCCCCN(C)C
さらに表示
Color scale=<30 APHA
Titration with HClO4>=96.0 %
Appearance (Form)Clear to slightly hazy liquid
Infrared spectrumConforms
Refractive index1.4233 to 1.4253 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only