Dodecyl sulfate, sodium salt, 85%, pure
Dodecyl sulfate, sodium salt, 85%, pure
Dodecyl sulfate, sodium salt, 85%, pure
Thermo Scientific Chemicals

Dodecyl sulfate, sodium salt, 85%, pure

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
製品番号(カタログ番号)数量パッケージ
226141000100 gPlastic Bottle
製品番号(カタログ番号) 226141000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
さらに表示
Appearance (Form)Powder or crystals
GC>=80 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only