meso-2,3-Dimercaptosuccinic acid, 98%
meso-2,3-Dimercaptosuccinic acid, 98%
meso-2,3-Dimercaptosuccinic acid, 98%
Thermo Scientific Chemicals

meso-2,3-Dimercaptosuccinic acid, 98%

CAS: 304-55-2 | C4H6O4S2 | 182.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 226671000
Price (EUR)
885,65
Online exclusive
984,00
Save 98,35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
885,65
Online exclusive
984,00
Save 98,35 (10%)
Each
Chemical Identifiers
CAS304-55-2
IUPAC Name2,3-disulfanylbutanedioic acid
Molecular FormulaC4H6O4S2
InChI KeyACTRVOBWPAIOHC-UHFFFAOYNA-N
SMILESOC(=O)C(S)C(S)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only