2'-Deoxyguanosine hydrate, 99+%
2'-Deoxyguanosine hydrate, 99+%
2'-Deoxyguanosine hydrate, 99+%
Thermo Scientific Chemicals

2'-Deoxyguanosine hydrate, 99+%

CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 226730010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS207121-55-9
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
UV spectrophotometrymolar absorptivity 13700 +/- 4%, on dry basis
Appearance (Form)Crystalline powder
Water=<8.0 % (K.F.)
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only