Methyl behenate, 99%, analytical standard for GC
Methyl behenate, 99%, analytical standard for GC
Methyl behenate, 99%, analytical standard for GC
Thermo Scientific Chemicals

Methyl behenate, 99%, analytical standard for GC

CAS: 929-77-1 | C23H46O2 | 354.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 226810010
Price (EUR)
55,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
55,10
Each
Specifications
Assay Percent Range98.5% min. (GC)
CAS929-77-1
Chemical Name or MaterialMethyl behenate
EINECS Number213-207-8
Formula Weight354.61
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only