Citric acid, trisodium salt dihydrate, 99%, pure
Citric acid, trisodium salt dihydrate, 99%, pure
Citric acid, trisodium salt dihydrate, 99%, pure
Thermo Scientific Chemicals

Citric acid, trisodium salt dihydrate, 99%, pure

CAS: 1545801 | C6H9Na3O9 | 294.1 g/mol
数量:
500 g
1 kg
5 kg
25 kg
パッケージ:
すべて表示
Fibre drum
製品番号(カタログ番号) 227130250
価格(JPY)
-
数量:
25 kg
パッケージ:
Fibre drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6132-04-3
IUPAC Nametrisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular FormulaC6H9Na3O9
InChI KeyNLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILESO.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
さらに表示
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Water11 to 13 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only