Hexadecyltrimethylammonium Bromide, 99+%
Hexadecyltrimethylammonium Bromide, 99+%
Hexadecyltrimethylammonium Bromide, 99+%
Thermo Scientific Chemicals

Hexadecyltrimethylammonium Bromide, 99+%

CAS: 57-09-0 | C19H42BrN | 364.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 227160101
Price (USD)
2,268.65
Online exclusive
2,521.00
Save 252.35
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
2,268.65
Online exclusive
2,521.00
Save 252.35
Each
Chemical Identifiers
CAS57-09-0
IUPAC Namehexadecyltrimethylazanium bromide
Molecular FormulaC19H42BrN
InChI KeyLZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(10% solution in water, after heating at 70°C)
SolubilityClear to slightly hazy, colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only