L(-)-Mannose, 99+%
L(-)-Mannose, 99+%
L(-)-Mannose, 99+%
Thermo Scientific Chemicals

L(-)-Mannose, 99+%

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number 227590010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10030-80-5
IUPAC Name(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
Molecular FormulaC6H12O6
InChI KeyGZCGUPFRVQAUEE-BXKVDMCESA-N
SMILESOC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
Loss on drying=<0.5 %
Heavy metals (as Pb)=<10 ppm
Infrared spectrumConforms
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only