5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals

5-Bromo-2'-deoxyuridine, 99+%

CAS: 59-14-3 | C9H11BrN2O5 | 307.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
2.5 g
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 228590010
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS59-14-3
IUPAC Name5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11BrN2O5
InChI KeyWOVKYSAHUYNSMH-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Br)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Water=<2 % (K.F.)
Residual solvents=<0.2 % (GC) Ethanol
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only