L-Methionine methyl ester hydrochloride, 98%
L-Methionine methyl ester hydrochloride, 98%
L-Methionine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Methionine methyl ester hydrochloride, 98%

CAS: 2491-18-1 | C6H14ClNO2S | 199.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
Packaging:
Glass bottle
Catalog number 229350250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2491-18-1
IUPAC Namehydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular FormulaC6H14ClNO2S
InChI KeyMEVUPUNLVKELNV-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC(=O)C(N)CCSC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 %
Solubility(5 % in methanol) Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Color)White to beige
Specific optical rotation+24° to +28° (20°C, 589 nm) (c=1, water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only