Myristyltrimethylammonium bromide, 99%
Myristyltrimethylammonium bromide, 99%
Myristyltrimethylammonium bromide, 99%
Thermo Scientific Chemicals

Myristyltrimethylammonium bromide, 99%

CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 229370050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1119-97-7
IUPAC Nametrimethyl(tetradecyl)azanium bromide
Molecular FormulaC17H38BrN
InChI KeyCXRFDZFCGOPDTD-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point245°C to 250°C
Appearance (Color)White
Titration Argentometric98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only