Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Polyvinylpyrrolidone-iodine complex
Thermo Scientific Chemicals

Polyvinylpyrrolidone-iodine complex

CAS: 25655-41-8 | (C6H9NO)nI2 | 364.95 g/mol
Have Questions?
Catalog NumberQuantity
229471000100 g
Catalog number 229471000
Price (USD)
99.65
Special offer
Online exclusive
Ends: 28-Dec-2026
117.00
Save 17.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 28-Dec-2026
117.00
Save 17.35
Each
Chemical Identifiers
CAS25655-41-8
Molecular Formula(C6H9NO)nI2
SMILESII.*-CC(-*)N1CCCC1=O
MDL NumberMFCD00084483
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to red-brown
Infrared spectrumConforms
Assay(on dried substance)
Sulfated ash=<0.025 % (2 g)
Loss on drying=<8 % (5 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only