2,3-Dicyanohydroquinone, 98%
2,3-Dicyanohydroquinone, 98%
2,3-Dicyanohydroquinone, 98%
Thermo Scientific Chemicals

2,3-Dicyanohydroquinone, 98%

CAS: 4733-50-0 | C8H4N2O2 | 160.132 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 232050250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4733-50-0
IUPAC Name3,6-dihydroxybenzene-1,2-dicarbonitrile
Molecular FormulaC8H4N2O2
InChI KeyMPAIWVOBMLSHQA-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C(C#N)=C1C#N
さらに表示
HPLC>=97.5 %
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Appearance (Color)Brown to khaki
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only