Formic acid-d2, for NMR, 99+ atom % D, 95% solution in D2O
Formic acid-d2, for NMR, 99+ atom % D, 95% solution in D2O
Formic acid-d2, for NMR, 99+ atom % D, 95% solution in D2O
Thermo Scientific Chemicals

Formic acid-d2, for NMR, 99+ atom % D, 95% solution in D2O

CAS: 920-42-3 | CH2O2 | 48.037 g/mol
Have Questions?
Catalog NumberQuantityPackaging
23340010010 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 233400100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS920-42-3
IUPAC Name(²H)methan(²H)oic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-PFUFQJKNSA-N
SMILES[2H]OC([2H])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration with NaOH>=94.0 %
Appearance (Form)Liquid
D-Enrichment>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only