Prostaglandin E2, 98%
Prostaglandin E2, 98%
Prostaglandin E2, 98%
Thermo Scientific Chemicals

Prostaglandin E2, 98%

CAS: 363-24-6 | C20H32O5 | 352.47 g/mol
Have Questions?
Catalog NumberQuantityPackaging
23356010010 mgGlass bottle
Catalog number 233560100
Price (USD)
530.65
Special offer
Online exclusive
Ends: 28-Dec-2026
624.00
Save 93.35
Each
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
530.65
Special offer
Online exclusive
Ends: 28-Dec-2026
624.00
Save 93.35
Each
Chemical Identifiers
CAS363-24-6
IUPAC Name(5E)-7-{3-hydroxy-2-[(1E)-3-hydroxyoct-1-en-1-yl]-5-oxocyclopentyl}hept-5-enoic acid
Molecular FormulaC20H32O5
InChI KeyXEYBRNLFEZDVAW-RTYMFESYNA-N
SMILESCCCCCC(O)\C=C\C1C(O)CC(=O)C1C\C=C\CCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
HPLC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only