L-Buthionine-(S,R)-sulfoximine, 99%
L-Buthionine-(S,R)-sulfoximine, 99%
L-Buthionine-(S,R)-sulfoximine, 99%
Thermo Scientific Chemicals

L-Buthionine-(S,R)-sulfoximine, 99%

CAS: 83730-53-4 | C8H18N2O3S | 222.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 235520010
Price (USD)
328.65
Online exclusive
365.00
Save 36.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
328.65
Online exclusive
365.00
Save 36.35
Each
Chemical Identifiers
CAS83730-53-4
IUPAC Name(2S)-2-azaniumyl-4-[butyl(imino)oxo-λ⁶-sulfanyl]butanoate
Molecular FormulaC8H18N2O3S
InChI KeyKJQFBVYMGADDTQ-CVSPRKDYSA-N
SMILESCCCCS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Specific optical rotation+31° to +35° (20°C, 589 nm) (c=1, 1 N HCl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only