Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Thermo Scientific Chemicals

Silica gel, for chromatography, 0.035-0.070 mm, 60 A

For column chromatography. | CAS: 7631-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
30 kg
Catalog number 240360010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH6.5 to 7.5 (5 % suspension in water) Typical
Particle size=<10 % (Undersize)
Residual water=<6 % (160°C, 15 min)
Appearance (Color)White
Specific gravity0.46 to 0.55 kg/l (Typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only