Methyl 3-oxo-pentanoate, 99+%
Methyl 3-oxo-pentanoate, 99+%
Methyl 3-oxo-pentanoate, 99+%
Thermo Scientific Chemicals

Methyl 3-oxo-pentanoate, 99+%

CAS: 30414-53-0 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 243165000
Price (USD)
449.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
449.00
Each
Chemical Identifiers
CAS30414-53-0
IUPAC Namemethyl 3-oxopentanoate
Molecular FormulaC6H10O3
InChI KeyXJMIXEAZMCTAGH-UHFFFAOYSA-N
SMILESCCC(=O)CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC=<0.5 % Methyl acetoacetate
GC>=99.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only