2,4,6-Trimethylbenzoyl chloride, 98%
2,4,6-Trimethylbenzoyl chloride, 98%
2,4,6-Trimethylbenzoyl chloride, 98%
Thermo Scientific Chemicals

2,4,6-Trimethylbenzoyl chloride, 98%

CAS: 938-18-1 | C10H11ClO | 182.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 244280500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS938-18-1
IUPAC Name2,4,6-trimethylbenzoyl chloride
Molecular FormulaC10H11ClO
InChI KeyUKRQMDIFLKHCRO-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(Cl)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5280 to 1.5300 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only