Propyl chloroformate, 98%
Propyl chloroformate, 98%
Propyl chloroformate, 98%
Thermo Scientific Chemicals

Propyl chloroformate, 98%

CAS: 109-61-5 | C4H7ClO2 | 122.55 g/mol
製品番号(カタログ番号) 245291000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-61-5
IUPAC Namepropyl carbonochloridate
Molecular FormulaC4H7ClO2
InChI KeyQQKDTTWZXHEGAQ-UHFFFAOYSA-N
SMILESCCCOC(Cl)=O
さらに表示
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4030 to 1.4050 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only