Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Magnesium sulfate heptahydrate, 99.0-100.5% (by anhydrous basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets BP, EP and USP requirements.
数量:
500 mL
2.5 L
40 lb.
450 lb.
パッケージ:
すべて表示
Clear glass bottle
製品番号(カタログ番号) 2504-14
価格(JPY)
-
数量:
500 mL
パッケージ:
Clear glass bottle
This product is GMP Manufactured and meets BP, EP and USP requirements.
This product is GMP Manufactured and meets BP, EP and USP requirements.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
仕様
CAS10034-99-8
化学物質名または材質Magnesium sulfate heptahydrate
Colorless
融点1124°C
分子式MgSO4·7H2O
パッケージClear glass bottle
異名Epsom salts
形状Crystalline Powder
分子量(g/mol)246.48
純度99 to 100.5%
数量500 mL
pH5 to 9.2 (25°C, 5% aqueous solution)
Unit SizeEach