Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™
Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™
Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™
Thermo Scientific Chemicals

Sodium tert-pentoxide, 30% in 2-MeTHF, AcroSeal™

CAS: 14593-46-5 | C5H11NaO | 110.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 252538000
Price (USD)
-
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS14593-46-5
IUPAC Namesodium 2,2-dimethylpropan-1-olate
Molecular FormulaC5H11NaO
InChI KeyYQMLGVNRTAQUFQ-UHFFFAOYSA-N
SMILES[Na+].CC(C)(C)C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
Titration with HClO4>=30 %
Appearance (Color)Yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only