4-Methylvaleric acid, 99%
4-Methylvaleric acid, 99%
4-Methylvaleric acid, 99%
Thermo Scientific Chemicals

4-Methylvaleric acid, 99%

CAS: 646-07-1 | C6H12O2 | 116.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2555100505 mLGlass bottle
Catalog number 255510050
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.00
Save 5.35
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.00
Save 5.35
Each
Chemical Identifiers
CAS646-07-1
IUPAC Name4-methylpentanoic acid
Molecular FormulaC6H12O2
InChI KeyFGKJLKRYENPLQH-UHFFFAOYSA-N
SMILESCC(C)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4136 to 1.4156 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only