5-Phenyltetrazole, 99%
5-Phenyltetrazole, 99%
5-Phenyltetrazole, 99%
Thermo Scientific Chemicals

5-Phenyltetrazole, 99%

CAS: 18039-42-4 | C7H6N4 | 146.15 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 263721000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18039-42-4
IUPAC Name5-phenyl-2H-1,2,3,4-tetrazol-2-ide
Molecular FormulaC7H5N4
InChI KeyLBCIMHAWIRLWHX-UHFFFAOYSA-N
SMILES[N-]1N=NC(=N1)C1=CC=CC=C1
さらに表示
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only